Learn More
Medchemexpress LLC Tylvalosin tartrate (acetylisovaleryltylosin tartrate) | 63428-13-7 | 96.03% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY128423500MG
Encompass_Preferred
Tylvalosin tartrate is an orally active, broad-spectrum macrolide antibiotic with antimicrobial activity. It acts as an antiviral agent useful in studying PRRSV infection and induces apoptosis. Additionally, it possesses anti-inflammatory activity, relieves oxidative stress, and alleviates acute lung injury by inhibiting NF-κB activation.
  • Orally active, broad-spectrum macrolide antibiotic
  • Antimicrobial activity
  • Antiviral agent for studying PRRSV infection
  • Induces apoptosis
  • Anti-inflammatory activity
  • Relieves oxidative stress
  • Alleviates acute lung injury by inhibiting NF-κB activation
Catalog No. 50-003-41203