Learn More
Cayman Chemical Tylvalosin (tartrate), Purity: ≥80%, CAS: 63428-13-7, 1 g
SDP
Catalog No. 502949945
Encompass_Preferred
A macrolide antibiotic; active against S. aureus, E. coli, and P. multocida (MICs = 2, 128, and 128 µg/ml, respectively); reduces LPS-induced production of pro-inflammatory cytokines, prostaglandin E2 (PGE2; Item No. 14010), and nitric oxide in RAW 264.7 cells at 5 and 10 µg/ml; reduces the severity of lung lesions in pigs infected with M. hyopneumoniae and P. multocida at 10 mg/kg per day in the drinking water
Catalog No. 50-294-9945 Supplier Cayman Chemical Supplier No. 338491 G
A macrolide antibiotic; active against S. aureus, E. coli, and P. multocida (MICs = 2, 128, and 128 µg/ml, respectively); reduces LPS-induced production of pro-inflammatory cytokines, prostaglandin E2 (PGE2; Item No. 14010), and nitric oxide in RAW 264.7 cells at 5 and 10 µg/ml; reduces the severity of lung lesions in pigs infected with M. hyopneumoniae and P. multocida at 10 mg/kg per day in the drinking water