Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 1890MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: P-GF.44C. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HUVEC cells. Breast, Bladder, Lung or Kaposi tumors. Cellular Staining: Cytoplasmic and Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human Thymidine Phosphorylase (TP / PD-ECGF) protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 55kDa.Chromosome: 22q13.33. Gene Symbol: TYMP. Gene ID: 1890. Ensemble: ENSG00000025708. Uniprot: NP_001107227.
Catalog No. 50-188-4654