Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7298MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: TS106 + TMS715. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: 5-FU-resistant colon carcinoma cell lines (NCI H630R10, NCI H630R1); 5-FU-resistant breast cancer cell lines, MCF-Ad5, MCF-Ad10. Testicular carcinomas. Cellular Staining: Nuclear & Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human thymidylate synthase (TS106 & TMS715). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 36kDa.Chromosome: 18p11.32. Gene Symbol: TYMS. Gene ID: 7298. Ensemble: ENSG00000176890. Uniprot: NP_001341796.
Catalog No. 50-188-5253