Learn More
Electron Microscopy Sciences Cargille Microscope Type 37 Immersion Oil 4 OZ
SDP
Supplier:  Electron Microscopy Sciences 1690904
Encompass_Preferred
Cargille PCB-Free Immersion oils were introduced in 1972. Today, no other immersion oil meets FDA criteria.  Cargille 37 Immersion Oils have an index of 1.515 at 37 deg C. Consideration of these oils is also suggested where stage temperatures are elevated by sub-stage illuminators and high wattage projection equipment.
Density: @ 23°C: 0.917 g/cc
Catalog No. 50-266-45