Learn More
IMEB, Inc Tek-Select Type 4 Cassettes with Attached Hinged Lid, Green, 1000 per Case, Easy Frosted Writing Surface
SDP
Supplier:  IMEB, Inc CG441
Encompass
Tek-Select Type 4 Cassettes offer a special hinge holding the base and cover together. This hinge allows cassettes to be opened and closed as often as necessary. The lid can be removed and re-inserted easily without danger of specimen loss. Three frosted writing surfaces are easy to write on, including the 45 deg. angle anterior face. Cassettes are packaged 1000 per case, with lids attached. Our best selling cassette!
Catalog No. 50-168-9511