Learn More
Perkin Elmer US LLC Type A0.5 MEINHARD Quartz Nebulizer for NexION 300/350
Supplier:  Perkin Elmer US LLC N8145011
Encompass_Preferred
MEINHARD Nebulizers are self-aspirating and not to be used with solutions containing hydrofluoric acid. Nebulizers require an end cap (N0680504) for use with Scott Spray Chambers.
Catalog No. 50-210-4814