Learn More
Invivogen ODN 2395 - TLR9 ligand, 5 mg
SDP
Supplier:  Invivogen TLRL23955
Encompass
CpG ODNs are synthetic oligonucleotides that contain unmethylated CpG dinucleotides in particular sequence contexts (CpG motifs).
Catalog No. NC0349514