Learn More
Advanced Biomatrix PureCol, Bovine Collagen, Lyophilized, 15 mg
Supplier:  Advanced Biomatrix 500615MG
Encompass_Preferred
The PureCol lyophilized product comes with 15 mg of type I bovine atelocollagen used for 2D coatings or as a collagen standard. PureCol is known as the standard among collagen products for purity (≥99.9% collagen content), consistency and reproducibility. PureCol is isolated from bovine hides sourced from the only controlled, closed herd in the United States. PureCol collagen is approximately 97% Type I with the remainder being comprised of Type III collagen. It is soluble atelocollagen. This product is supplied as a sterile lyophilized powder with 15 mg of bovine collagen. When reconstituted with 5 ml of sterile 0.01 N HCl, a concentration of approximately 3 mg/ml is achieved. PureCol collagen is ideal for coating of surfaces and providing preparations of thin layers for culturing cells. PureCol, lyophilized form is not recommended for the formation of a solid gel. PureCol collagen is provided in user-friendly packaging for use and storage.
Catalog No. 50-487-4