Learn More
Cole-Parmer Type-K High-Temperature Fiberglass-Insulated Probe Mini Conn Exposed 10ft 20-Gauge
Supplier:  Cole-Parmer 0846724
Encompass_Preferred
Digi-Sense Type-K High-Temperature Fiberglass-Insulated Probe Mini-Connector, Exposed 10ft 20 -Gauge, High-Temperature Fiberglass Insulated Wire Used for High-Temperature MeasurementsResponse Time: 0.75 seconds or greaterAll insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type K - yellow, and type J – black..Custom designs or changes to this product are available upon request, -Fiberglass wire is not recommended for use in moist or wet environments. -Exposed-junction probes are not recommended for use with aqueous solutions. -Do not subject type J exposed-junction probes to moisture as they will rust.
Catalog No. NC1102989