Learn More
TARGETMOL CHEMICALS INC TYPHANEOSIDE 25MG
SDP
Catalog No. 502580116
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg and bulk. Please contact Fisher for quotes. Typhaneoside (Aervitrin) is a flavonoid glycoside plant extract with potential ability to treat primary dysmenorrhea through the Shaofu Zhuyu decoction. purity: 99%
Catalog No. 50-258-0116 Supplier TARGETMOL CHEMICALS INC Supplier No. T380025MG
Also available in 5 mg 10 mg 50 mg and bulk. Please contact Fisher for quotes. Typhaneoside (Aervitrin) is a flavonoid glycoside plant extract with potential ability to treat primary dysmenorrhea through the Shaofu Zhuyu decoction. purity: 99%