Learn More
Creative Biomart Recombinant SARS MERS Spike Glycoprotein S1 Antigen, Fc-tagged
Catalog No. 502085116
Encompass_Preferred
The HEK293 derived recombinant protein contains the SARS MERS Spike Glycoprotein S1, amino acids 18-725 fused to Fc tag at C-terminal.
Catalog No. 50-208-5116 Supplier Creative Biomart Supplier No. VAC0276
The HEK293 derived recombinant protein contains the SARS MERS Spike Glycoprotein S1, amino acids 18-725 fused to Fc tag at C-terminal.