Learn More
CPC Scientific H-Tyr-Ser-Phe-Val-His-His-Gly-Phe-Phe-Asn-Phe-Arg-Val-Ser-Trp-Arg-Glu-Met-Leu-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP054B
Encompass
Peptide that binds an epitope on the cyclin-dependent kinase 2 surface with a Kd ~ 38 nM. Furthermore, it inhibits Cdk2 activity while serving as a monoclonal antibody analog.ONE-LETTER SEQUENCE: YSFVHHGFFNFRVSWREMLAMOLECULAR FORMULA: C121H164N32O27S1MOLECULAR WEIGHT:2530.86STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [175799-54-9], RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: P. Colas et al., Nature 380, 548 (1996)
Catalog No. 50-211-0213