Learn More
CPC Scientific H-Tyr-Ser-Pro-Lys-Met-Val-Gln-Gly-Ser-Gly-Cys-Phe-Gly-Arg-Lys-Met-Asp-Arg-Leu-Ser-Ser-Ser-Ser-Gly-Leu-Gly-Cys-Lys-Val-Leu-Arg-Arg-His-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NATR012B
Encompass
ONE-LETTER SEQUENCE: YSPKMVQGSGCFGRKMDRLSSSSGLGCKVLRRH (Cys11 and 27 bridge)MOLECULAR FORMULA: C152H253N51O44S4MOLECULAR WEIGHT:3627.26STORAGE CONDITIONS: -20 5CSYNONYMS: (Tyr0)-Brain Natriuretic Peptide-32 (human), ventricular natriuretic peptide, (Tyr)-BNP-32 (human)RESEARCH AREA: Cardiovascular
Catalog No. 50-210-9863