Learn More
CPC Scientific H-Tyr-Ala-Cys-Asp-Thr-Ala-Thr-Cys-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asn-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OSTP019B
Encompass
Calcitonin gene related peptide (CGRP) is part of the calcitonin family of peptides, which in humans exists in two forms, alpha-CGRP and beta-CGRP. alpha-CGRP is a 37-amino acid peptide formed from the alternative genetic splicing of the calcitonin/CGRP geneONE-LETTER SEQUENCE: YACDTATCVTHRLAGLLSRSGGVVKNNFVPTNVGSKAF-NH2 (Cys3 and 8 bridge)MOLECULAR FORMULA: C172H276N52O51S2MOLECULAR WEIGHT:3952.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [124756-98-5], SYNONYMS: Tyr-alpha-CGRP (human), Tyr-CGRP I (human)RESEARCH AREA: CardiovascularREFERENCES: J Biol Chem. 2006 Mar 17, 281(11):7205-13
Catalog No. 50-211-0034