Learn More
CPC Scientific H-Tyr-His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP002B
Encompass
ONE-LETTER SEQUENCE: YHKTDSFVGLM-NH2MOLECULAR FORMULA: C59H89N15O16S1MOLECULAR WEIGHT:1296.51STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: 116868-93-0, SYNONYMS: (Tyr)-Neurokinin ARESEARCH AREA: Neuropeptides
Catalog No. 50-210-9883