Learn More
CPC Scientific Ac-Tyr-DPhe-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110610
Encompass
VIP receptor antagonist.ONE-LETTER SEQUENCE: Ac-Y-DPhe-DAIFTNSYRKVLGQLSARKLLQDIMSR-NH2MOLECULAR FORMULA: C157H252N44O43S1MOLECULAR WEIGHT:3476.02STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [93965-89-0], SYNONYMS: Acetyl-(D-Phe2)-GRF (1-29) amide (human), Acetyl-(D-Phe)-SermorelinRESEARCH AREA: GastrointestinalREFERENCES: Weelbroeck et al., Endocrinology, 116, 2643 (1985)
Catalog No. 50-211-0610 Supplier CPC Scientific Supplier No. VIP002A
VIP receptor antagonist.ONE-LETTER SEQUENCE: Ac-Y-DPhe-DAIFTNSYRKVLGQLSARKLLQDIMSR-NH2MOLECULAR FORMULA: C157H252N44O43S1MOLECULAR WEIGHT:3476.02STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [93965-89-0], SYNONYMS: Acetyl-(D-Phe2)-GRF (1-29) amide (human), Acetyl-(D-Phe)-SermorelinRESEARCH AREA: GastrointestinalREFERENCES: Weelbroeck et al., Endocrinology, 116, 2643 (1985)