Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Tyr36)-pTH-Related Protein (1-36) (human, mouse, rat) | 213779-11-4 | 99.1% | 4309.85 | 10 MG
SDP

Catalog No. 5000398590
Encompass_Preferred

(Tyr36)-pTH-Related Protein (1-36) (human, mouse, rat) is a peptide that functions as a parathyroid hormone (PTH) receptor ligand. It is intended for research use only.

  • White to off-white solid
  • Purity: 99.1%
  • CAS Number: 213779-11-4
  • Molecular weight: 4309.85
  • Molecular formula: C194H303N59O53
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-98590 Supplier Medchemexpress LLC Supplier No. HYP471510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Tyr36)-pTH-Related Protein (1-36) (human, mouse, rat) is a peptide that functions as a parathyroid hormone (PTH) receptor ligand. It is intended for research use only.

  • White to off-white solid
  • Purity: 99.1%
  • CAS Number: 213779-11-4
  • Molecular weight: 4309.85
  • Molecular formula: C194H303N59O53
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.