Learn More
Biotium Ready-to-Use Tyramide Amplification Buffer, 1X, 25ML
Catalog No. 501964403
Encompass_Preferred
Ready-to-use tyramide amplification buffer can be used for tyramide signal amplification with CF Dye Tyramides or other tyramides without the need to add hydrogen peroxide.
Catalog No. 50-196-4403 Supplier Biotium Supplier No. 2202725ML
Ready-to-use tyramide amplification buffer can be used for tyramide signal amplification with CF Dye Tyramides or other tyramides without the need to add hydrogen peroxide.