Learn More
Biotium Tyramides and Tyramide Signal Amplification Kits Fluorescence Microscopy Reactive CF Dyes Other Dyes Biotinylation Reagents TyraMax 630 650 1/EA
Supplier:  Biotium 9614020UL
Encompass_Preferred
TyraMax Dyes are next-gen tyramide (TSA) dyes for spatial biology with colors from blue to near-IR named for their excitation/emission They are stable in amplification buffer with advantages in color selection brightness and photostability vs other TSA dyes 100X in DMSO 20 uL trial size
Catalog No. 50-005-01444