Learn More
Biotium Tyramides and Tyramide Signal Amplification Kits Fluorescence Microscopy Reactive CF Dyes Other Dyes Biotinylation Reagents TyraMax 630 650 1/EA
Catalog No. 5000501482
Encompass_Preferred
TyraMax Dyes are next-generation tyramide (TSA) dyes for spatial biology with colors from blue to near-IR named for their excitation/emission They are stable in amplification buffer with advantages in color selection brightness and photostability versus other TSA dyes 100X in DMSO 100 uL
Catalog No. 50-005-01482 Supplier Biotium Supplier No. 96140100UL
TyraMax Dyes are next-generation tyramide (TSA) dyes for spatial biology with colors from blue to near-IR named for their excitation/emission They are stable in amplification buffer with advantages in color selection brightness and photostability versus other TSA dyes 100X in DMSO 100 uL