Learn More
Cayman Chemical Tyramine-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 1189884-47-6, 5 mg
SDP
Catalog No. 502941234
Encompass_Preferred
An internal standard for the quantification of tyramine by GC- or LC-MS
Catalog No. 50-294-1234 Supplier Cayman Chemical Supplier No. 285315 MG
An internal standard for the quantification of tyramine by GC- or LC-MS