Learn More
Medchemexpress LLC HY-W007606 1g Medchemexpress, Tyramine CAS:51-67-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0076061G
Encompass_Preferred
Medchemexpress, HY-W007606 1g Tyramine CAS:51-67-2 Tyramine is an indirectly acting sympathomimetic amine that releases norepinephrine from presynaptic nerve terminals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4497