Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tyrannophos

Catalog No. 9022841G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-228-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-228-41G Supplier Sigma Aldrich Supplier No. 9022841G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This new aryl alkylphosphine ligand developed in Abby's Doyle's lab has been tailored specifically for nickel catalysis as demonstrated in the Suzuki coupling of acetals with boronic acids to generate benzylic ethers, reactions that pose challenges in Ni catalysis when using phosphine ligands developed for Pd-catalysed couplings.Parameterization of phosphine ligands demonstrates enhacement of nickel catalysis via remote steric effects

Specifications

CAS 2138440-82-9
Melting Point 292°C
Molecular Formula C5H4NNaO2S
Linear Formula C5H4NNaO2S
Quantity 1 g
Synonym 2-Pyridinesulfinic acid; Sodium salt; Willis 2-pyridylsulfinates H
Molecular Weight (g/mol) 165.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.