Learn More
TYRELL MEDICAL LLC Tyrell Chemical HTN-130 AIR
Catalog No. 502734059
Encompass_Preferred
Inert dielectric high performance HFE thermal transfer fluid, 20kg/drum Sea freight. Heat condution of machinery and equipment, electronics and datacenter cooling, immersion computer cooling. CAS 29770-93-9. 130C Boil Point
Catalog No. 50-273-4059 Supplier TYRELL MEDICAL LLC Supplier No. HTN1305KGSEA
Inert dielectric high performance HFE thermal transfer fluid, 20kg/drum Sea freight. Heat condution of machinery and equipment, electronics and datacenter cooling, immersion computer cooling. CAS 29770-93-9. 130C Boil Point