Learn More
ACROBiosystems Recombinant Protein;Human TYRO3 / Dtk Protein, Fc Tag;HEK293;1MG;TY3,TYRO3,BYK,DTK,RSE,SKY,TIF,TYRO-3
Catalog No. 501658532
Encompass_Preferred
Recombinant Protein;Human TYRO3 / Dtk Protein, Fc Tag;1MG;Human TYRO3, Fc Tag (TY3-H5251) is expressed from human 293 cells (HEK293). It contains AA Ala 41 - Ser 428 (Accession # AAH51756).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68.2 kDa. The protein migrates as 85-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8532 Supplier ACROBiosystems Supplier No. TY3H52511MG
Recombinant Protein;Human TYRO3 / Dtk Protein, Fc Tag;1MG;Human TYRO3, Fc Tag (TY3-H5251) is expressed from human 293 cells (HEK293). It contains AA Ala 41 - Ser 428 (Accession # AAH51756).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68.2 kDa. The protein migrates as 85-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.