Learn More
Boston Bioproducts Inc Tyrode's Solution - 500ml
Supplier:  Boston Bioproducts Inc BSS340500ML
Encompass
Tyrode’s Balanced Salt Solution (TBSS) is an isotonic buffer solution that plays a crucial role in cell culture and tissue preservation. It is formulated with inorganic salts and a carbohydrate energy source to provide cells with essential nutrients while maintaining a stable pH and osmotic balance, which is vital for optimal cell growth and functionality.The balanced composition of TBSS maintains the structural and physiological integrity of cells during biological research and development.
Catalog No. NC2299120