Learn More
Boston Bioproducts Inc Tyrode's Solution (with HEPES & 0.25% BSA) - 500ml
Catalog No. NC2012638
Encompass
Tyrode’s Solution is an isotonic buffer solution that plays a crucial role in cell culture and tissue preservation. It is formulated with inorganic salts, HEPES, BSA, and a carbohydrate energy source to provide cells with essential nutrients while maintaining a stable pH and osmotic balance. The balanced composition of this buffer maintains the structural and physiological integrity of cells.
Catalog No. NC2012638 Supplier Boston Bioproducts Inc Supplier No. BSS370500ML
Tyrode’s Solution is an isotonic buffer solution that plays a crucial role in cell culture and tissue preservation. It is formulated with inorganic salts, HEPES, BSA, and a carbohydrate energy source to provide cells with essential nutrients while maintaining a stable pH and osmotic balance. The balanced composition of this buffer maintains the structural and physiological integrity of cells.