Learn More
Boston Bioproducts Inc Tyrode's Solution (with HEPES, with Calcium) - 1L
Catalog No. NC2267202
Encompass
Tyrode’s modified I Solution is an isotonic buffer solution that plays a crucial role in cell culture and tissue preservation. It is formulated with inorganic salts and a carbohydrate energy source to provide cells with essential nutrients while maintaining a stable pH and osmotic balance, which is vital for optimal cell growth and functionality. The balanced composition of Tyrodes maintains the structural and physiological integrity of cells during biological research and development.
This item is not returnable. View return policy
Catalog No. NC2267202 Supplier Boston Bioproducts Inc Supplier No. BSS3551L
Tyrode’s modified I Solution is an isotonic buffer solution that plays a crucial role in cell culture and tissue preservation. It is formulated with inorganic salts and a carbohydrate energy source to provide cells with essential nutrients while maintaining a stable pH and osmotic balance, which is vital for optimal cell growth and functionality. The balanced composition of Tyrodes maintains the structural and physiological integrity of cells during biological research and development.