Learn More
Sigma Aldrich Fine Chemicals Biosciences Tyrode's Salts With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | 1L
Catalog No. NC1984023
Encompass
Tyrode's Salts With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | 1L
Catalog No. NC1984023 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T23971L
Tyrode's Salts With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | 1L