Learn More
Medchemexpress LLC HY-106263B 10mg Medchemexpress, Tyroserleutide (hydrochloride) CAS:852982-42-4 Purity:>98%
SDP
Catalog No. 501968333
Encompass_Preferred
Medchemexpress, HY-106263B 10mg Tyroserleutide (HCl) CAS:852982-42-4 Tyroserleutide hydrochloride, isolated from the degradation products of porcine spleen, is a small molecular tripeptide which inhibits tumor growth both in vitro and in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8333 Supplier Medchemexpress LLC Supplier No. HY106263B10MG
Medchemexpress, HY-106263B 10mg Tyroserleutide (HCl) CAS:852982-42-4 Tyroserleutide hydrochloride, isolated from the degradation products of porcine spleen, is a small molecular tripeptide which inhibits tumor growth both in vitro and in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.