Learn More
Sigma Aldrich Fine Chemicals Biosciences Tyrosinase from mushroom, 9002-10-2, MFCD00082118, 25 KU
Catalog No. NC0563226
Encompass
Lyophilized powder, ≥1000 unit/mg solid, Synonym: Catechol Oxidase, Monophenol Monooxygenase, Monophenol, dihydroxyphenylalanine:oxygen oxidoreductase, Polyphenol Oxidase. Tyrosinase activity has been assessed in a study that developed an alternative therapeutic agent for treating hyperpigmentation. Tyrosinase has also been used in a study to investigate the oculocutaneous albinism phenotype in the Pakistani population.
Catalog No. NC0563226 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T382425KU
Lyophilized powder, ≥1000 unit/mg solid, Synonym: Catechol Oxidase, Monophenol Monooxygenase, Monophenol, dihydroxyphenylalanine:oxygen oxidoreductase, Polyphenol Oxidase. Tyrosinase activity has been assessed in a study that developed an alternative therapeutic agent for treating hyperpigmentation. Tyrosinase has also been used in a study to investigate the oculocutaneous albinism phenotype in the Pakistani population.