Learn More
Enzo Life Sciences Tyrosinase (human), (recombinant) (His-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5350100
Encompass_Preferred
Alternative name: Monophenol monooxygenase. Purity: ≥85% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS/HCl, pH 8.0, 1M NaCl, 0.05% Empigen BB, and 30% glycerol. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2074