Learn More
CPC Scientific H-Ser-Glu-Ile-Trp-Arg-Asp-Ile-Asp-Phe-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PKCP052A
Encompass
Antigenic peptide presented by HLA-B44 that is recognized on human melanoma by autologous cytolytic Tlymphocytes. ONE-LETTER SEQUENCE: SEIWRDIDFMOLECULAR FORMULA: C54H77N13O17MOLECULAR WEIGHT:1180.28STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [170294-35-6], SYNONYMS: Tyrosinase (192-200) (human, mouse)RESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0208