Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrosine, 2,5-dihydroxy- | 21373-30-8 | 95.2% | 213.19 g·mol⁻¹ | C9H11NO5 | 5 MG
SDP

Catalog No. 5000264217
Encompass_Preferred

6-Hydroxy-DOPA is a small-molecule research compound reported as a selective, allosteric inhibitor of the RAD52 ssDNA-binding domain and is used in cancer research. It is supplied as a solid in small quantities and is characterized by its chemical composition and analytical data.

  • Selective allosteric inhibitor of the RAD52 ssDNA-binding domain.
  • Suitable for cancer research and biochemical assay studies.
  • Solid, off-white to gray appearance for standard handling and storage.
  • Molecular formula C9H11NO5; molecular weight 213.19 g·mol⁻¹.
  • Purity (HPLC) 95.2% as reported on the COA.
  • Store powder at -20°C (stable for up to 3 years); in solution store at -80°C for extended stability.
  • Soluble in DMSO at approximately 50 mg/mL; ultrasonic agitation recommended for dissolution.

Catalog No. 50-002-64217 Supplier Medchemexpress LLC Supplier No. HY1102865MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Hydroxy-DOPA is a small-molecule research compound reported as a selective, allosteric inhibitor of the RAD52 ssDNA-binding domain and is used in cancer research. It is supplied as a solid in small quantities and is characterized by its chemical composition and analytical data.

  • Selective allosteric inhibitor of the RAD52 ssDNA-binding domain.
  • Suitable for cancer research and biochemical assay studies.
  • Solid, off-white to gray appearance for standard handling and storage.
  • Molecular formula C9H11NO5; molecular weight 213.19 g·mol⁻¹.
  • Purity (HPLC) 95.2% as reported on the COA.
  • Store powder at -20°C (stable for up to 3 years); in solution store at -80°C for extended stability.
  • Soluble in DMSO at approximately 50 mg/mL; ultrasonic agitation recommended for dissolution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.