Learn More
Medchemexpress LLC Tyrosine Hydroxylase Antibody (YA023) | 59 kDa | 100 UL
SDP
Catalog No. 5000397188
Encompass_Preferred
Tyrosine Hydroxylase Antibody (YA023) is a high-quality, rabbit-derived monoclonal antibody. This non-conjugated IgG antibody specifically targets Tyrosine Hydroxylase and exhibits reactivity across human, mouse, and rat species. It is provided in a solution containing TBS (pH 7.4), BSA, and Glycerol, with Sodium Azide as a preservative, ensuring stability and longevity. The antibody is suitable for a wide range of applications in molecular biology and immunology research.
  • Specifically targets Tyrosine Hydroxylase
  • Reactive with human, mouse, and rat samples
  • Suitable for Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), Flow Cytometry (FC), and Immunofluorescence-Tissue (IF-Tissue) applications
  • Supplied as a stable liquid formulation
  • Recommended storage at -20°C for optimal preservation
Catalog No. 50-003-97188 Supplier Medchemexpress LLC Supplier No. HYP80362100UL
Tyrosine Hydroxylase Antibody (YA023) is a high-quality, rabbit-derived monoclonal antibody. This non-conjugated IgG antibody specifically targets Tyrosine Hydroxylase and exhibits reactivity across human, mouse, and rat species. It is provided in a solution containing TBS (pH 7.4), BSA, and Glycerol, with Sodium Azide as a preservative, ensuring stability and longevity. The antibody is suitable for a wide range of applications in molecular biology and immunology research.
  • Specifically targets Tyrosine Hydroxylase
  • Reactive with human, mouse, and rat samples
  • Suitable for Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), Flow Cytometry (FC), and Immunofluorescence-Tissue (IF-Tissue) applications
  • Supplied as a stable liquid formulation
  • Recommended storage at -20°C for optimal preservation