Learn More
Enzo Life Sciences Tyrosine hydroxylase polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA4970100
Encompass_Preferred
Immunogen: Denatured tyrosine hydroxylase (TH) from rat pheochromocytoma. Species reactivity: Mammalian. Applications: ICC, IHC, WB. Purity: Affinity purified. Formulation: Liquid. In 10mM HEPES, pH 7.5, containing 150mM NaCl, 10% BSA and 50% glycerol.
Catalog No. 50-201-1840