Learn More
Medchemexpress LLC HY-100315 10mg Medchemexpress, Tyrosine kinase-IN-1 CAS:705946-27-6 Purity:>98%
SDP
Catalog No. 501931776
Encompass_Preferred
Medchemexpress, HY-100315 10mg Tyrosine kinase-IN-1 CAS:705946-27-6 Tyrosine kinase-IN-1 is a multi-targeted tyrosine kinase inhibitor with IC 50 s of 4, 20, 4, 2 nM for KDR , Flt-1 , FGFR1 and PDGFRα , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1776 Supplier Medchemexpress LLC Supplier No. HY10031510MG
Medchemexpress, HY-100315 10mg Tyrosine kinase-IN-1 CAS:705946-27-6 Tyrosine kinase-IN-1 is a multi-targeted tyrosine kinase inhibitor with IC 50 s of 4, 20, 4, 2 nM for KDR , Flt-1 , FGFR1 and PDGFRα , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.