Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrosine kinase-in-7 | 345615-74-9 | 99.3% | 297.37 g/mol | C16H15N3OS | 10 MG
SDP

Catalog No. 5000287809
Encompass_Preferred

Tyrosine kinase-IN-7 is a small-molecule research compound that inhibits the epidermal growth factor receptor (EGFR) tyrosine kinase. It is supplied as a solid with molecular formula C16H15N3OS and molecular weight 297.37 g/mol; handle and store according to the product SDS.

  • Small-molecule epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor.
  • Solid form suitable for dissolution and in vitro assays.
  • Molecular formula C16H15N3OS.
  • Molecular weight 297.37 g/mol.
  • Reported purity approximately 99.25%.
  • Storage recommendations include refrigerated storage at 4°C; solutions may be stored at -20°C or -80°C per SDS.

Catalog No. 50-002-87809 Supplier Medchemexpress LLC Supplier No. HY15691210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrosine kinase-IN-7 is a small-molecule research compound that inhibits the epidermal growth factor receptor (EGFR) tyrosine kinase. It is supplied as a solid with molecular formula C16H15N3OS and molecular weight 297.37 g/mol; handle and store according to the product SDS.

  • Small-molecule epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor.
  • Solid form suitable for dissolution and in vitro assays.
  • Molecular formula C16H15N3OS.
  • Molecular weight 297.37 g/mol.
  • Reported purity approximately 99.25%.
  • Storage recommendations include refrigerated storage at 4°C; solutions may be stored at -20°C or -80°C per SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.