Learn More
Medchemexpress LLC Tyrosine kinase inhibitor | 1021950-26-4 | MFCD17010276 | 99.9% | 586.61 g/mol | C31H31FN6O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1042125MG
Encompass_Preferred
Tyrosine kinase inhibitor is a research-grade small-molecule inhibitor (CAS 1021950-26-4) used to probe protein tyrosine kinase and receptor tyrosine kinase signaling in biochemical and cellular studies. It is supplied as a white to off-white solid with high reported purity and comes in multiple pack sizes and solution formats to support assay development and screening.
  • High purity (≈99.9%).
  • Molecular weight 586.61 g/mol.
  • White to off-white solid appearance.
  • Available as solid (5-100 mg) and 10 mM solutions in DMSO.
  • Suitable for biochemical and cellular research applications.
  • Stable when stored under recommended conditions for powder and in solvent.
Catalog No. 50-002-43496