Learn More
Medchemexpress LLC Tyrosine kinase inhibitor | 34823-86-4 | 99.9% | 234.25 | 1 ML
SDP
Catalog No. 5000344076
Encompass_Preferred
GTP-14564 is a tyrosine kinase inhibitor targeting internal tandem duplication (ITD) and FLT3, which inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells.
  • Targets internal tandem duplication (ITD) and FLT3.
  • Inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells.
  • Used in cancer targeted therapy.
  • Functions as an inhibitor of Fms-like tyrosine kinase 3 (FLT3).
Catalog No. 50-003-44076 Supplier Medchemexpress LLC Supplier No. HY110026100MG
GTP-14564 is a tyrosine kinase inhibitor targeting internal tandem duplication (ITD) and FLT3, which inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells.
  • Targets internal tandem duplication (ITD) and FLT3.
  • Inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells.
  • Used in cancer targeted therapy.
  • Functions as an inhibitor of Fms-like tyrosine kinase 3 (FLT3).