Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrosine, N-[(2E)-3-(4'-chloro[1,1'-biphenyl]-4-yl)-1-oxo-2-propen-1-yl]-O-(phenylmethyl)- | 907952-06-1 | 99.3% | C31H26ClNO4 | 10MG
SDP

Catalog No. 5000258959
Encompass_Preferred

GPR34 receptor antagonist 2 is a small-molecule antagonist of the G protein-coupled receptor GPR34 provided for preclinical research. It is intended for studies of immune and inflammatory disease mechanisms and for investigation of histamine-release modulation in mast cells.

  • High purity (99.26%).
  • Molecular formula C31H26ClNO4.
  • Molecular weight 512.00 g/mol.
  • Available as solid and as 10 mM solution in DMSO.
  • Supplied in multiple milligram package sizes suitable for assay development.
  • Used for immune and inflammatory disease research and histamine-release studies.

Catalog No. 50-002-58959 Supplier Medchemexpress LLC Supplier No. HY13850110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GPR34 receptor antagonist 2 is a small-molecule antagonist of the G protein-coupled receptor GPR34 provided for preclinical research. It is intended for studies of immune and inflammatory disease mechanisms and for investigation of histamine-release modulation in mast cells.

  • High purity (99.26%).
  • Molecular formula C31H26ClNO4.
  • Molecular weight 512.00 g/mol.
  • Available as solid and as 10 mM solution in DMSO.
  • Supplied in multiple milligram package sizes suitable for assay development.
  • Used for immune and inflammatory disease research and histamine-release studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.