Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrosylleucine | 17355-10-1 | 99.9% | 25 MG
SDP

Catalog No. 5000230422
Encompass_Preferred

Tyrosylleucine (Tyr-Leu, YL) is an orally active dipeptide reported to exhibit potent antidepressant-like activity. It is supplied as a high-purity research-grade compound for preclinical in vitro and in vivo studies.

  • Sequence Tyr-Leu dipeptide
  • Chemical formula C15H22N2O4, molecular weight 294.35 g·mol⁻¹
  • High purity (99.9% by HPLC)
  • Supplied in milligram quantities suitable for research use
  • Stable as a solid when stored under recommended conditions

Catalog No. 50-002-30422 Supplier Medchemexpress LLC Supplier No. HY12279425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrosylleucine (Tyr-Leu, YL) is an orally active dipeptide reported to exhibit potent antidepressant-like activity. It is supplied as a high-purity research-grade compound for preclinical in vitro and in vivo studies.

  • Sequence Tyr-Leu dipeptide
  • Chemical formula C15H22N2O4, molecular weight 294.35 g·mol⁻¹
  • High purity (99.9% by HPLC)
  • Supplied in milligram quantities suitable for research use
  • Stable as a solid when stored under recommended conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.