Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrosylleucine | 17355-10-1 | 99.9% | 50 MG
SDP

Catalog No. 5000230757
Encompass_Preferred

Tyrosylleucine (Tyr-Leu, YL) is an orally active dipeptide composed of tyrosine and leucine, reported to exhibit potent antidepressant-like activity. It is supplied as a high-purity white to off-white solid for research use, with specified storage and handling recommendations.

  • Sequence Tyr-Leu (dipeptide).
  • Cas number 17355-10-1.
  • Molecular weight 294.35 g/mol.
  • Purity 99.9% (as stated by supplier).
  • Appearance white to off-white solid.
  • Storage sealed and protected from moisture; powder: -80°C for 2 years, -20°C for 1 year.
  • Available in milligram pack sizes, including 50 mg.

Catalog No. 50-002-30757 Supplier Medchemexpress LLC Supplier No. HY12279450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrosylleucine (Tyr-Leu, YL) is an orally active dipeptide composed of tyrosine and leucine, reported to exhibit potent antidepressant-like activity. It is supplied as a high-purity white to off-white solid for research use, with specified storage and handling recommendations.

  • Sequence Tyr-Leu (dipeptide).
  • Cas number 17355-10-1.
  • Molecular weight 294.35 g/mol.
  • Purity 99.9% (as stated by supplier).
  • Appearance white to off-white solid.
  • Storage sealed and protected from moisture; powder: -80°C for 2 years, -20°C for 1 year.
  • Available in milligram pack sizes, including 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.