Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7306MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM611. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: SK-MEL-23, SK-MEL-19, SK-MEL-30, SK-MEL-37 cells or Melanoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human TYRP1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 75kDa.Chromosome: 9p23. Gene Symbol: TYRP1. Gene ID: 7306. Ensemble: ENSG00000107165. Uniprot: NP_000541.
Catalog No. 50-188-5259