Learn More
Aviva Systems Biology Corp TYRP1 Antibody - N-terminal region (ARP44313_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44313_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TYRP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TYRP1. Alternate names: CAS2, CATB, GP75, TRP, TYRP, b-PROTEINApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53415