Learn More
Enzo Life Sciences Tyrphostin 23 (RG-50810) (10mg). CAS: 118409-57-7
SDP
Catalog No. 502010599
Encompass_Preferred
A potent inhibitor of EGF receptor kinase activity (IC50=35 µM). Alternative name: alpha-Cyano-(3,4-dihydroxy)cinnamonitrile. Purity: ≥99% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0599 Supplier Enzo Life Sciences Supplier No. BMLEI1910010
A potent inhibitor of EGF receptor kinase activity (IC50=35 µM). Alternative name: alpha-Cyano-(3,4-dihydroxy)cinnamonitrile. Purity: ≥99% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: Ambient.