Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin 25 | 118409-58-8 | MFCD00133900 | 99.0% | 202.17 | C10H6N2O3 | 10 MG
SDP

Catalog No. 5000245851
Encompass_Preferred

Tyrphostin 25 (AG82) is a small-molecule research compound that selectively inhibits epidermal growth factor receptor (EGFR) tyrosine kinase and acts as a GPR35 agonist. It prevents EGFR autophosphorylation and reduces EGF-induced cell migration in vitro. Supplied as a yellow to brown solid with high analytical purity for laboratory research; for research use only.

  • High purity: 99.0%.
  • Molecular weight: 202.17 g/mol.
  • Chemical formula: C10H6N2O3.
  • Physical form: solid, yellow to brown powder.
  • Biological activity: EGFR IC50 ≈ 3 μM (A431 cells); GPR35 EC50 ≈ 5.3 μM.
  • Storage: powder -20°C (stable for years); in solution -80°C (months).
  • SDS available in multiple languages.
  • Intended use: for research use only.

Catalog No. 50-002-45851 Supplier Medchemexpress LLC Supplier No. HY10195810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrphostin 25 (AG82) is a small-molecule research compound that selectively inhibits epidermal growth factor receptor (EGFR) tyrosine kinase and acts as a GPR35 agonist. It prevents EGFR autophosphorylation and reduces EGF-induced cell migration in vitro. Supplied as a yellow to brown solid with high analytical purity for laboratory research; for research use only.

  • High purity: 99.0%.
  • Molecular weight: 202.17 g/mol.
  • Chemical formula: C10H6N2O3.
  • Physical form: solid, yellow to brown powder.
  • Biological activity: EGFR IC50 ≈ 3 μM (A431 cells); GPR35 EC50 ≈ 5.3 μM.
  • Storage: powder -20°C (stable for years); in solution -80°C (months).
  • SDS available in multiple languages.
  • Intended use: for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.