Learn More
Enzo Life Sciences Tyrphostin 47 (RG-50864) (AG-213) (5mg). CAS: 122520-86-9
SDP
Catalog No. 502010595
Encompass_Preferred
A potent inhibitor of EGF receptor kinase activity (IC50‘s:EGFR = 2.4 µM, PDGFR = 3.5 µM, p210bcr-abl=5.9 µM, insulin receptor = 640 µM). Purity: ≥99% (TLC). Solubility: Soluble at 20 mg/ml in 100% ethanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-0595 Supplier Enzo Life Sciences Supplier No. BMLEI1880005
A potent inhibitor of EGF receptor kinase activity (IC50‘s:EGFR = 2.4 µM, PDGFR = 3.5 µM, p210bcr-abl=5.9 µM, insulin receptor = 640 µM). Purity: ≥99% (TLC). Solubility: Soluble at 20 mg/ml in 100% ethanol or DMSO. Long Term Storage: -20°C.