Learn More
Enzo Life Sciences Tyrphostin 8 (10mg). CAS: 3785-90-8
SDP
Catalog No. 502011509
Encompass_Preferred
The tyrphostins are well known tyrosine kinase inhibitors. However, tyrphostin 8 has been shown to inhibit calcineurin (IC50=21 µM) while displaying weak inhibition of EGFR tyrosine kinase (IC50=560 µM). Alternative name: alpha-Cyano-(4-hydroxy)cinnamonitrile. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (25mg/ml) or DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1509 Supplier Enzo Life Sciences Supplier No. BMLPR1130010
The tyrphostins are well known tyrosine kinase inhibitors. However, tyrphostin 8 has been shown to inhibit calcineurin (IC50=21 µM) while displaying weak inhibition of EGFR tyrosine kinase (IC50=560 µM). Alternative name: alpha-Cyano-(4-hydroxy)cinnamonitrile. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (25mg/ml) or DMSO (25mg/ml). Long Term Storage: Ambient.